EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Correlation of nuclear color and opalescence with protein S-thiolation in human lenses.
PMID 10328968
Abstract
Human lens nuclei were collected during routine cataract surgery and used to study the role of oxidation in cataract formation and brunescence. This study focused on the comparison of the intensities of nuclear opacity and pigmentation (brunescence) with the changes in free glutathione (GSH) and the three species of protein-thiol mixed disulfides: protein-S-S-glutathione (PSSG), protein-S S-cysteine (PSSC) and protein-S-S-gamma-glutamylcysteine (PSSGC). Eighty-one freshly excised human lens nuclei from a population with a mean age of 77 were used. The nuclear color was graded using the CCRG system, ranging from yellow to dark brown. The nuclear cataract opalescence of these lenses was also graded using the LOCS II system, ranging from LOCS II NO-1 to NO-4. Three normal human lenses (average age of 88 yr) were also included in the study as controls. The nuclear samples were each analyzed for free GSH and protein-thiol mixed disulfides, respectively. It was found that nuclear GSH decreased as the nuclear color increased from yellow to dark brown (from 0.73+/-0.13 to 0.13+/-0.03 micromole g wet wt-1) and as the nuclear opalescence increased from NO.1 to NO.4 (from 0. 80+/-0.19 to 0.20+/-0.01 micromole g wet wt-1). All these values were lower than that of GSH in normal controls (1.43+/-0.59 micromole g wet wt-1). Levels of both PSSG and PSSC progressively increased, however, as the nuclear color intensified. PSSG increased from 0.29+/-0.05 to 0.91+/-0.11 micromole g wet wt-1while PSSC increased from 0.13+/-0.04 to 0.41+/- 0.06 micromole g wet wt-1. PSSGC concentration progressively increased with increases in both nuclear pigmentation (from 0.05+/-0.01 to 0.23+/-0.05 micromole g wet wt-1) and nuclear opacity (from 0.02+/-0.00 to 0.20+/-0.02 micromole g wet wt-1). In comparison, normal controls had lower levels of all three mixed disulfide species: PSSG, 0.22+/-0.06; PSSC, 0.08+/-0.02; PSSGC, 0.02+/-0.06 micromole g wet wt-1, respectively. The correlation of lens nuclear color and opalescence intensity with nuclear protein S-thiolation indicates that protein-thiol mixed disulfides may play an important role in cataractogenesis and development of brunescence in human lenses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0903
γ-Glu-Cys, ≥80% (HPLC)
C8H14N2O5S