EMAIL THIS PAGE TO A FRIEND
Natural product reports
Thiazole and oxazole peptides: biosynthesis and molecular machinery.
PMID 10331285
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151645
Thiazole, 99%
C3H3NS