EMAIL THIS PAGE TO A FRIEND
Pharmacology, biochemistry, and behavior
The effects of intravenous naltrindole and naltrindole 5'-isothiocyanate on sufentanil-induced respiratory depression and antinociception in rats.
PMID 10340539
Abstract
Although the interactions between the mu- and the delta-opiate receptor subtypes are well documented with regard to supraspinal analgesia, less is known about the mutual interactions on respiratory depression. To clarify the functional interactions between both opiate receptor subtypes with regard to antinociception and respiratory depression, male Wistar rats were intravenously injected with 2.5 microg/kg of the mu-opiate agonist sufentanil and subsequently intravenously challenged with the delta antagonist naltrindole (NTI) or naltrindole 5'-isothiocyanate (5'-NTII), a delta-2 antagonist. Antinociception was measured by means of the tail-flick latency, and respiratory depression was evaluated by means of analysis of PaCO2, PaO2, and oxygen saturation. To quantify the antagonistic properties of NTI and 5'-NTII, mean areas under the curve were calculated for groups treated with sufentanil, control vehicle, and sufentanil plus a dose of the antagonists. NTI, but not 5'-NTII, antagonized the sufentanil-induced antinociception at 10 mg/kg NTI. Below this dose the effects were inconsistent. The sufentanil-induced hypercapnia and hypoxia were diminished with 10 mg/kg NTI or 5'-NTII. These data indicate that NTI antagonizes the sufentanil-induced antinociception and respiratory depression in rats. A dissociation between the antinociception and respiratory depression following intravenous sufentanil could be obtained with 10 mg/kg 5'-NTII pointing to different regulatory effects of opiate delta receptor subtypes on mu-opiate agonist-induced behavioral effects.