EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
Isomeric and anomeric specificity of the metabolic and secretory response of rat pancreatic islets to glucose pentaacetate.
PMID 10341285
Abstract
In the presence of 2.8 mM D-glucose, beta-D-glucose pentaacetate (1. 7 mM) augmented insulin release from isolated rat pancreatic islets more than alpha-D-glucose pentaacetate. Likewise, the further increment in insulin output evoked by nateglinide (0.01 mM) was higher in islets exposed to beta- rather than alpha-D-glucose pentaacetate. Inversely, in the presence of 2.8 mM unesterified D-glucose, alpha-L-glucose pentaacetate, but not beta-L-glucose pentaacetate, significantly augmented insulin output. The higher insulinotropic potency of the beta-anomer of D-glucose pentaacetate coincided with the fact that it significantly increased the paired ratio between D-[U-14C]glucose oxidation and D-[5-3H]glucose utilization, whereas alpha-D-glucose pentaacetate failed to do so. These findings are interpreted to support the concept that the stimulation of insulin release by these esters is largely attributable to their direct interaction with a stereospecific receptor, with preference for the configuration of the C1 common to beta-D-glucose pentaacetate and alpha-L-glucose pentaacetate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2354
α-D(+)-Glucose pentaacetate, 99%
C16H22O11
285943
β-D-Glucose pentaacetate, 98%
C16H22O11