EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Malarone (atovaquone and proguanil hydrochloride): a review of its clinical development for treatment of malaria. Malarone Clinical Trials Study Group.
PMID 10348225
Abstract
The continuing spread of drug-resistant malaria emphasizes the need for new antimalarial drugs. Atovaquone is a broad-spectrum antiprotozoal drug with a novel mechanism of action, via inhibition of parasite mitochondrial electron transport, and a favorable safety profile. Early studies with atovaquone alone for treatment of malaria demonstrated good initial control of parasitemia but an unacceptable rate of recrudescent parasitemia. Parasites isolated during recrudescence after treatment with atovaquone alone were resistant to atovaquone in vitro. The combination of atovaquone and proguanil is synergistic in vitro, and clinical studies demonstrated enhanced efficacy of the combination compared to either drug alone for treatment of malaria. Malarone, a fixed-dose combination of 250 mg of atovaquone and 100 mg of proguanil hydrochloride, is available in many countries for treatment of acute, uncomplicated malaria caused by Plasmodium falciparum. At the recommended dose (in adults, four tablets once a day for three days), the overall cure rate was > 98% in more than 500 patients with falciparum malaria. In four randomized, controlled clinical trials, treatment with atovaquone and proguanil hydrochloride was significantly more effective than mefloquine (Thailand), amodiaquine (Gabon), chloroquine (Peru and the Philippines) or chloroquine plus pyrimethamine/sulfadoxine (Philippines). In clinical trials where the comparator drug was highly effective, treatment with atovaquone and proguanil hydrochloride was equally effective. Parasites isolated during recrudescence after treatment with the combination of atovaquone and proguanil were not resistant to atovaquone in vitro. The most commonly reported adverse events in clinical trials (abdominal pain, anorexia, nausea, vomiting, diarrhea and coughing) occurred with similar frequency in patients treated with a comparator drug. Malarone is a safe and effective new agent for treatment of malaria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7048
Proguanil hydrochloride, ≥95% (HPLC)
C11H16ClN5·HCl