EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Asymmetric synthesis of L-azetidine-2-carboxylic acid and 3-substituted congeners--conformationally constrained analogs of phenylalanine, naphthylalanine, and leucine.
PMID 10360752
Abstract
Enantiopure L-azetidine-2-carboxylic acid, the (3R)-phenyl, (3R)-naphthyl and (3S)-isopropyl analogs were prepared based on a zinc-mediated asymmetric addition of allylic halides to the camphor sultam derivative of glyoxylic acid O-benzyl oxime.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5637
3-(1-Naphthyl)-DL-alanine
C13H13NO2