EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Antichymotrypsin interaction with chymotrypsin. Intermediates on the way to inhibited complex formation.
PMID 10364215
Abstract
Serpins form enzymatically inactive covalent complexes (designated E*I*) with their target proteinases, corresponding most likely to the acyl enzyme that resembles the normal intermediate in substrate turnover. Formation of E*I* involves large changes in the conformation of the reactive center loop (residues P17 to P9') and of the serpin molecule in general. The "hinge" region of the reactive center loop, including residues P10-P14, shows facile movement in and out of beta-sheet A, and this movement appears to be crucial in determining whether E*I* is formed (the inhibitor pathway) or whether I is rapidly hydrolyzed to I* (the substrate pathway). Here, we report stopped-flow and rapid quench studies investigating the pH dependence of the conversion of the alpha1-antichymotrypsin.alpha-chymotrypsin encounter complex, E.I, to E*I*. These studies utilize fluorescent derivatives of cysteine variants of alpha1-antichymotrypsin at the P11 and P13 residues. Our results demonstrate three identifiable intermediates, EIa, EIb, and EIc, between E.I and E*I* and permit informed speculation regarding the nature of these intermediates. Partitioning between inhibitor and substrate pathways occurs late in the process of E*I* formation, most likely from a species occurring between EIc and E*I*.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3639
4-Fluoro-7-sulfamoylbenzofurazan
C6H4FN3O3S