EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Identification of a general transcription factor TFIIAalpha/beta homolog selectively expressed in testis.
PMID 10364255
Abstract
In this paper we describe the isolation of a cDNA that encodes a human TFIIAalpha/beta-like factor (ALF). The open reading frame of ALF predicts a protein of 478 amino acids that contains characteristic N- and C-terminal conserved domains separated by an internal nonconserved domain. In addition, a rare ALF-containing cDNA, which possesses an extended N terminus (Stoned B/TFIIAalpha/beta-like factor; SALF) has also been identified. The results of Northern and dot blot analyses show that ALF is expressed almost exclusively in testis; in contrast, TFIIAalpha/beta and TFIIAgamma are enriched in testis but are also widely expressed in other human tissues. Recombinant ALF (69 kDa) and TFIIAgamma (12 kDa) polypeptides produced in Escherichia coli form an ALF/gamma complex that can stabilize TBP-TATA interactions in an electrophoretic mobility shift assay. The ALF/gamma complex is also able to restore transcription from the adenovirus major late promoter using HeLa cell nuclear extracts that have been depleted of TFIIA. Overall, the data show that ALF is a functional homolog of human general transcription factor TFIIAalpha/beta that may be uniquely important to testis biology.