EMAIL THIS PAGE TO A FRIEND
Experimental Cell Research
NA22598, a novel antitumor compound, reduces cyclin D1 levels, arrests cell cycle at G1 phase, and inhibits anchorage-independent growth of human tumor cells.
PMID 10366423