The Biochemical journal
Intramolecular chaperone and inhibitor activities of a propeptide from a bacterial zinc aminopeptidase.
PMID 10377241
An aminopeptidase from Aeromonas caviae T-64 was translated as a preproprotein consisting of three domains; a signal peptide (19 amino acid residues), an N-terminal propeptide (101 residues) and a mature region (273 residues). We demonstrated that a proteinase, which was isolated from the culture filtrate of A. caviae T-64, activated the recombinant pro-aminopeptidase by removal of the majority of the propeptide. Using L-Leu-p-nitroanilide as a substrate, the processed aminopeptidase showed a large increase in kcat when compared with the unprocessed enzyme, whereas the Km value remained relatively unchanged. The similar Km values for the pro-aminopeptidase and the mature aminopeptidase indicated that the N-terminal propeptide of the pro-aminopeptidase did not influence the formation of the enzyme-substrate complex, suggesting the absence of marked conformational changes in the active domain. In contrast, the marked difference in kcat suggests a significant decrease in the energy of one or more of the transition states of the enzyme-substrate reaction coordinate. Moreover, we showed that the activity of the urea-denatured pro-aminopeptidase could be recovered by dialysis, whereas the activity of the urea-denatured mature aminopeptidase, which lacked the propeptide, could not. Further to this, the propeptide-deleted aminopeptidase formed an inclusion body in the cytoplasmic space in Escherichia coli and was not secreted at all. These results suggested that the propeptide of the pro-aminopeptidase acted as an intramolecular chaperone that was involved with the correct folding of the enzyme in vitro and was required for extracellular secretion in E. coli.