EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
What is the origin of 3-methylglutaconic acid?
PMID 10384380
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M47604
3-Methylglutaric acid, 99%
C6H10O4