EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Biochemical and neurotoxicological effects of L-2-chloropropionic acid on rodent brain.
PMID 10386989
Abstract
L-2-Chloropropionic acid (L-CPA) is selectively toxic to cerebellar granule cells; necrosis is first observed in rats 36 h after L-CPA administration (750 mg/kg p.o.) and becomes marked by 48 h. L-CPA has also been shown to activate the mitochondrial pyruvate dehydrogenase (PDH) complex in fasted adult rats, resulting in reduced blood glucose and lactate levels. This study aimed to investigate the biochemical and neurotoxicological effects of L-CPA on the brain. Extracts, prepared from guinea-pig cerebellar and cerebral cortex slices incubated in the presence of L-CPA, were analysed using 1H magnetic resonance spectroscopy, 31P magnetic resonance spectroscopy, and amino acid analysis. Glucose metabolism was studied by monitoring the metabolism of [1-(13)C]glucose using gas chromatography/mass spectrometry. Increased glucose metabolism and decreases in the pool sizes of lactate and alanine were observed in both tissues, demonstrating activation of the PDH complex. Extracts were also prepared from the forebrain and cerebellum of animals that had been treated in vivo with L-CPA and analysed as described for the in vitro studies. Similar evidence for PDH activation was demonstrated at 2 and 24 h after dosing in both tissues. At 48 h after dosing, when signs of toxicity are observed, an increase in the lactate concentration and a decrease in N-acetylaspartate in the cerebellum but not in the forebrain confirmed the selective neurotoxic action of L-CPA. These results suggest that activation of the PDH complex does not directly lead to the delayed selective neurotoxicity of L-CPA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109274
2-Chloropropionic acid, 92%
C3H5ClO2