EMAIL THIS PAGE TO A FRIEND
Vaccine
Effect of the bacillus of Calmette-Guérin, Propionibacter acnes and avridine as immunomodulators in antirabies vaccination of mice using the Fuenzalida-Palacios mouse brain vaccine.
PMID 10392627
Abstract
Using the laboratory mice, Fuenzalida-Palacios mouse brain human rabies vaccine was administered in groups of animals previously inoculated with rabies virus and then submitted to treatments with the immunomodulators onco-BCG, avridine and Propionibacterium acnes. Humoral and cellular immune responses were evaluated through the macrophage inhibition factor (MIF), intra-pad inoculation (IPI) and serum neutralization (SN) tests and by the detection of gamma-interferon (IFN-gamma). The IPI test was not effective in detecting the response of delayed-type hypersensitivity, contrary to MIF, which showed the immune cellular response. Higher levels of IFN-gamma were observed in the groups of mice vaccinated and treated with avridine and P. acnes. Although immunomodulating activities have been detected, the use of adjuvants with the Fuenzalida-Palacios type vaccine in mice did not reveal any encouraging results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0123
Avridine, ≥97% (HPLC)
C43H90N2O2