EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
A GC/MS/MS screening method for multiple organic acidemias from urine specimens.
PMID 10404733
Abstract
A gas chromatography tandem mass spectrometry method using an ion trap GC/MS system was developed to quickly screen urine samples for 14 organic acids associated with multiple organic acidemias. The following organic acids are used as diagnostic markers: methylmalonic acid, glutaric acid, 2-ketoisocaproic acid, succinylacetone, 3-methylcrotonylglycine, tiglylglycine, isovalerylglycine, fumaric acid, butyrylglycine, propionylglycine, hexanoylglycine, adipic acid, suberic acid, and sebacic acid. 2-ketocaproic acid is used as an internal standard. The samples are prepared using a solid-phase extraction and converted to trimethylsilyl derivatives. The extraction efficiency for the 14 compounds is between 57 and 106%. A derivatized standard mixture of the 14 markers is run prior to the patient samples to determine the accurate absolute and relative retention times. The samples are then injected and the product ion spectra monitored. For data analysis, one characteristic product ion plot is extracted for each of the 14 marker compounds, and the presence of a peak with the expected retention time is determined. The areas of the product ion peaks are compared with the reference range determined from 30 normal controls. Ten samples of patients with known organic acidemias were measured. For all patients, diagnostic peaks at the expected retention times of at least five times the upper limit of the reference range were detected. The method, with its relatively fast sample preparation, short 10.0 min run time and simple data analysis, is suitable for use as a quick metabolic screen of very sick patients in whom there is concern regarding the possibility of a treatable inborn error.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60930
Suberic acid, purum, ≥98.0% (T)
C8H14O4
S5200
Suberic acid, 98%
C8H14O4