EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Comparison of the antioxidant action of the alcoholic extract of Rubia cordifolia with rubiadin.
PMID 10410466
Abstract
The inhibition of FeSO4 induced lipid peroxidation in rat liver by alcoholic extract of Rubia cordifolia and by one of its constituent rubiadin (1, 3-dihydroxy-2-methyl anthraquinone) (pure form) has been compared. Both have been found to inhibit lipid peroxidation in a dose dependent manner. Whereas the former shows both oxidising and reducing properties with Fe2+ and Fe3+, the latter shows oxidising property only by converting Fe2+ to Fe3+. The former inhibits the oxidation of reduced glutathione while the latter does not.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74553
Rubiadin, analytical standard
C15H10O4