EMAIL THIS PAGE TO A FRIEND
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme
[Leptomycin: a specific inhibitor of protein nuclear export].
PMID 10412289
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6417
Leptomycin A, from Streptomyces sp., ~95% (HPLC), methanol solution
C32H46O6