EMAIL THIS PAGE TO A FRIEND
American journal of human genetics
The bare lymphocyte syndrome: molecular clues to the transcriptional regulation of major histocompatibility complex class II genes.
PMID 10417269