EMAIL THIS PAGE TO A FRIEND
American journal of therapeutics
Alprostadil: modes of actions in peripheral arterial occlusive disease.
PMID 10423631
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000054
Alprostadil, European Pharmacopoeia (EP) Reference Standard
C20H34O5