EMAIL THIS PAGE TO A FRIEND
International angiology : a journal of the International Union of Angiology
Neutrophil count and amputation in critical limb ischaemia.
PMID 10424370
Abstract
The white blood cell count (WCC) is known to be predictive of cardiac and cerebral vascular events. No one has yet investigated this in critical limb ischaemia (CLI). Baseline WCC was examined in relation to lower limb amputation 6 months after a 4 week treatment period with i.v. placebo or i.v. taprostene in 366 patients with CLI. The WCC was related to a significant increase in amputation, relative risk 1.6 (p=0.001, 95% CI: 1.2-2.0) in CLI patients with WCC > or =9x10(9)/l vs patients with WCC <9x10(9)/l. Its association with disabling amputation persisted on logistic regression analyses which included cigarette smoking as a variable and also treatment group (p<0.001). The WCC is therefore an easily measurable prognostic variable in CLI. The white blood cell may promote intractable tissue ischaemia by capillary plugging and/or release of toxic chemicals and may have distinct effects on tissue viability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4949
Taprostene sodium salt, ≥98% (HPLC)
C24H29NaO5