EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
The branched-chain dodecylbenzene sulfonate degradation pathway of Pseudomonas aeruginosa W51D involves a novel route for degradation of the surfactant lateral alkyl chain.
PMID 10427075
Abstract
Pseudomonas aeruginosa W51D is able to grow by using branched-chain dodecylbenzene sulfonates (B-DBS) or the terpenic alcohol citronellol as a sole source of carbon. A mutant derived from this strain (W51M1) is unable to degrade citronellol but still grows on B-DBS, showing that the citronellol degradation route is not the main pathway involved in the degradation of the surfactant alkyl moiety. The structures of the main B-DBS isomers and of some intermediates were identified by gas chromatography-mass spectrometric analysis, and a possible catabolic route is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44178
1-Phenyldodecane, analytical standard
C18H30
113239
1-Phenyldodecane, 97%
C18H30