EMAIL THIS PAGE TO A FRIEND
General pharmacology
Pharmacokinetic profile and adverse gastric effect of zinc-piroxicam in rats.
PMID 10428018
Abstract
Complexation of piroxicam with zinc extends its absorption time in rats. The time of peak concentration value for complexed piroxicam was 5.27 hr compared to only 2.56 hr for the uncomplexed agent. Piroxicam and zinc-piroxicam show similar inhibitory effects on carrageenin-induced paw edema. Zinc-piroxicam is less irritating than piroxicam on the gastric mucosa.