EMAIL THIS PAGE TO A FRIEND
Applied occupational and environmental hygiene
The occupational exposure level (OEL) for 2-bromopropane: the first OEL established by Korea.
PMID 10429728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B78114
2-Bromopropane, 99%
C3H7Br
59370
2-Bromopropane, purum, ≥99.0% (GC)
C3H7Br