EMAIL THIS PAGE TO A FRIEND
Cancer investigation
Ab initio studies of some amino acid residue complexes with 4-mercaptopyridine as a model for thymitaq (AG337), an inhibitor of thymidylate synthase.
PMID 10434950
Abstract
The complex formed by isopentane, as a model for the isoleucine residue present in the wild-type thymidylate synthase, with 4-mercaptopyridine as a fragment of the thymidylate synthase inhibitor Thymitaq (AG337) is investigated with ab initio quantum chemical calculations at Hartree-Fock and MP2 levels, using the 3-21G* basis set. The binding energy is compared with the binding energies of 4-mercaptopyridine with amino acid residues found in mutant thymidylate synthase enzymes. As compared with isoleucine, alanine and glycine do not show binding, in agreement with enzyme-inhibition results.