EMAIL THIS PAGE TO A FRIEND
The Analyst
Multi-residue analysis of tranquillizers in meat: confirmatory assays using mass spectrometry.
PMID 10435288
Abstract
A rapid and sensitive multi-residue method was developed to attempt to confirm the presence of the beta-blocker carazolol and the tranquillizers acepromazine, azaperone, chlorpromazine, propionylpromazine and xylazine in pig muscle tissues. The procedure involves determination by liquid chromatography coupled with tandem mass spectrometry. The liquid chromatographic separation was performed on a Symmetry C18 column with gradient elution. A mixture of aqueous buffer, containing 0.01% m/v trifluoroacetic acid (pH 3.5), and acetonitrile at a flow rate of 0.4 ml min-1 was used as the mobile phase. The abundant parent ions [M+ H+] produced by positive electrospray ionisation were selected for collisional dissociation with argon. Fragment ions were recorded with daughter ion scan and multiple reaction monitoring. The analytes were identified unambiguously by assessing retention times and diagnostic ions in meat samples spiked from 50 micrograms kg-1 [maximum residue limit (MRL) for azaperone and azaperol] to 5 micrograms kg-1 (MRL for carazolol).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7780
Propionylpromazine hydrochloride
C20H24N2OS · HCl
46689
Propionylpromazine hydrochloride, VETRANAL, analytical standard
C20H24N2OS · HCl