EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effect of an acaricide on the reproduction and survival of Daphnia magna.
PMID 10441632
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45680
Tetradifon, PESTANAL®, analytical standard
C12H6Cl4O2S