EMAIL THIS PAGE TO A FRIEND
Allergy
Sensitization to triglycidylisocyanurate (TGIC) with cutaneous and respiratory manifestations.
PMID 10442533
Abstract
The case is presented of a man with allergic contact dermatitis and occupational asthma due to triglycidylisocyanurate (TGIC), which is used as a hardener in thermosetting powder paint. The contact dermatitis was confirmed by patch testing (TGIC 0.5% and 5% in petrolatum), and the occupational asthma was confirmed by bronchial provocation testing: two challenges to an aerosol of lactose containing TGIC (0.05% and 0.1%, w/w, each for 0.5+1+2+4 min) led to a maximal decrease in FEV1 of 22% and 31% after 6 and 4 h, respectively. Skin prick tests with unconjugated TGIC were possibly positive. This case confirms that exposure to TGIC in powder paints may cause not only contact dermatitis, but also occupational asthma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379506
Tris(2,3-epoxypropyl) isocyanurate
C12H15N3O6