EMAIL THIS PAGE TO A FRIEND
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie
Participation of peroxidase reaction in thiamine catabolism.
PMID 10445402
Abstract
The horse-radish peroxidase catalyzes oxidation of thiamine in vitro in presence of hydroperoxide hydrogen (H2O2) and olenic acid (LOOH) (as an emultion) with formation of thiochrome (2,7-dimethyl-8-(oxyetyl-5,6-dihydrothiazolo (2,3-a) pyrimido (4,5-d) pyrimidine, oxodihydrothiochrome (2,7dimethyl-oxo-8-(-2-oxo-ethyl)-5,6,9a, 10-tetrahydrothiazolo (2,3-a) pyrimido (4,5-d) pyrimidine and thiamine disulfide (bis (3-methyl-((1-methyl-4-amino 5 pyrimidine methylformyl) amino 1-2 (oxyethyl)-1-propenyl) disulfide. The transformation of thiamine to thiochrome cause by release of two atoms of hydrogen from NH2-group of pyrimidine ring of molecule of thiamine and occurrence on this place of bond with thiozole ring. The microsomal fraction of rats in vitro in presence of LOOH also activities the oxidation of thiamine to thiochrome. However in these conditions peroxidase reaction is proceeded more slowly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7891
Thiochrome, analytical standard
C12H14N4OS