EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Interaction of cytocidal drugs and the inhibition of caspase-3 by 3-nitrosobenzamide.
PMID 10446456
Abstract
The effect of 3-nitrosobenzamide (NOBA) on the etoposide, staurosporine and dexamethason induced rapid (4-6 hr), caspase-dependent apoptosis was investigated in thymocytes and lymphoma cells by flow cytometric assay of DNA fragmentation. When NOBA (ED(50) = 4 microM) was added to these cell systems, the rapid onset of apoptosis was prevented. Such apparent protection by NOBA was related to the inactivation of caspase-3, by s-nitrosylation of 1.3 mol -SH per enzyme molecule out of 7 -SH groups. Since NOBA by itself induces DNA fragmentation within 18 hr in lymphoma cells, our results indicate that at least two active cell death pathways exist with apparent dissimilar kinetics and molecular mechanisms.