EMAIL THIS PAGE TO A FRIEND
Histochemistry and cell biology
Highly sensitive chelating agents for histochemical staining of rare earth metals.
PMID 10460472
Abstract
In tissues, bromopyridylazo-diethylaminophenol has been found to be capable of staining very small amounts of rare earth metals, particularly praseodymium, terbium, dysprosium, holmium, ytterbium, and lutetium. Differentiation of a target metal from interfering metals was achieved using masking agents, polyphosphates and aminopolycarboxylic acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
180017
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol, 97%
C15H17BrN4O