EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Exposure-pattern dermatitis due to 2-aminothiophenol and 2-aminophenyldisulfide.
PMID 10475529
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274240
2-Aminothiophenol, 99%
C6H7NS
123137
2-Aminothiophenol, technical grade, 90%
C6H7NS