EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
Allyl isovalerate.
PMID 10476390
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W204501
Allyl isovalerate, ≥98%, FCC
C8H14O2