EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
Lauroyl peroxide.
PMID 10476431
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290785
Luperox® LP, Lauroyl peroxide, 97%
C24H46O4