EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
Tris(2,3-dibromopropyl) phosphate.
PMID 10476477
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34188
TBPP, PESTANAL®, analytical standard
C9H15Br6O4P