EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
Vinyl bromide.
PMID 10476478
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1902
Vinyl bromide, 98%
C2H3Br
434191
Vinyl bromide solution, 1.0 M in THF
C2H3Br