EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Reporter cell lines to study the estrogenic effects of xenoestrogens.
PMID 10492897
Abstract
In order to characterize the estrogenic activity of chemicals, we established complementary in vitro recombinant receptor-reporter gene assays in stably transfected MCF-7 and HeLa cells. MCF-7 cells which express the endogenous estrogen receptor alpha (ER alpha) were stably transfected with only an estrogen-regulated luciferase gene. These cells enable the detection of compounds which bind to ER alpha or interfere with the induction of ER alpha mediated gene expression. Furthermore, HeLa cells, which do not express endogenous ERs, were transfected with an ER alpha or an ER beta construct together with an estrogen-regulated luciferase gene, or a chimeric GAL4-ER alpha receptor and the corresponding luciferase reporter gene. Finally, we tested these four cellular models as tools to check the estrogenic activities of several potential xenoestrogens and to detect estrogenic activity in wastewater sewage treatment effluents. In all of the models, nonylphenol mixture (NPm), 4n-nonylphenol (4nNP), 2,4'-DDE, 4,4'-DDE and wastewater sewage treatment effluent were active, while PCB mixture (Aroclor 1254), PCB 77, atrazine and lindane (gamma hexachlorocyclohexane) were inactive. Dioxin partially activates the estrogen receptor in MCF-7 cells while in HeLa-derived cell lines, it decreased the estrogenic-induced expression of luciferase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36663
2,4′-DDE solution, 100 μg/mL in methanol, PESTANAL®, analytical standard
C14H8Cl4