EMAIL THIS PAGE TO A FRIEND
Neuropsychobiology
Does a glutamate receptor blocker (Caroverine) enhance haloperidol in the treatment of schizophrenia?
PMID 10494049
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1119
Caroverine hydrochloride, ≥98% (HPLC), solid
C22H28ClN3O2