EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Residues of pesticides in grains locally grown in Saudi Arabia.
PMID 10501721
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241288
Triphenyl phosphate, ≥99%
C18H15O4P
442829
Triphenyl phosphate, analytical standard
C18H15O4P