EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Mobilization of trivalent chromium in presence of organic acids: a hydroponic study of wheat plant (Triticum vulgare).
PMID 10501732