EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Plasmodium berghei: induction of aminopeptidase in malaria-resistant strain of Anopheles gambiae.
PMID 10502473
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9125
L-Leucine-p-nitroanilide, leucine aminopeptidase substrate
C12H17N3O3