EMAIL THIS PAGE TO A FRIEND
Journal of investigative medicine : the official publication of the American Federation for Clinical Research
Phospholipase C activation by Na+/Ca2+ exchange is essential for monensin-induced Ca2+ influx and arachidonic acid release in FRTL-5 thyroid cells.
PMID 10510591
Abstract
Monensin, a Na+ ionophore, can increase cytosolic Ca2+ ([Ca2+]i) by reversing the Na+/Ca2+ exchange mechanism. This study provided additional insights into the mechanism of this Na+ ionophore-induced increase in [Ca2+]i, and emphasized the critical role of phospholipase C (PLC) in amplifying Na+/Ca2+ exchange-induced Ca2+ influx and subsequent arachidonic acid (AA) release in FRTL-5 thyroid cells. The possible involvement of protein kinase C (PKC), mitogen-activated protein kinase (MAPK), and GTP-binding (G) protein in mediating monensin-induced AA release was also explored. FRTL-5 thyroid cells were maintained in Coon's modified Ham's F-12 medium supplemented with a 6-hormone (6H) mixture. Cytosolic Ca2+ was measured by using indo-1 AM and a dual-wave-length spectrofluorometer. Release of 3H-labeled inositol trisphosphates and arachidonic acid were determined by a scintillation counter. In Hank's balanced salt solution with Ca2+ (HBSS+), monensin (100 mumol/L) induced a 2.3-fold sustained Ca2+ increase associated with IP3 generation and a 6-fold increase in AA release. Deletion of extracellular Ca2+, or replacement of Na+ by choline chloride in the medium, reduced the [Ca2+]i increase by 77% and completely prevented the monensin-induced rise in AA release. Similar inhibitory effects were observed in cells pretreated with a Na+ channel blocker, or Na+/Ca2+ exchange inhibitors. In HBSS without Ca2+ (HBSS-), monensin induced a 1-fold transient [Ca2+]i increase but did not increase the AA. This Ca2+ increase was not suppressed by U-73122, a PLC inhibitor. In HBSS+, U-73122 did not affect the monensin-induced initial transient peak increase of [Ca2+]i, but reduced the sustained second phase of [Ca2+]i from 400 nmol/L to 250 nmol/L, and completely blocked AA release. A phospholipase A2 (PLA2) inhibitor blocked the monensin-induced AA release without affecting the [Ca2+]i increase. Inhibition of PKC prevented 87% to 94% of the monesin-stimulated AA release. The monensin-induced AA release was also inhibited 94% by pertussis and 51% by a MAP kinase cascade inhibitor. The results suggest that monensin initiates an increase in [Ca2+]i via a Na+/Ca2+ exchange mechanism that triggers more pronounced and sustained [Ca2+]i increase via activation of PLC and Ca2+ influx. The PLC activation, followed by sustained Ca2+ influx and PKC activation, is a prerequisite for PLA2-mediated processes in monensin-challenged FRTL-5 thyroid cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5273
Monensin sodium salt, 90-95% (TLC)
C36H61NaO11