EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational allergic contact dermatitis from epoxy resins based on bisphenol F.
PMID 10515117
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B46808
Bis(2-hydroxyphenyl)methane, 98%
C13H12O2