EMAIL THIS PAGE TO A FRIEND
The Journal of eukaryotic microbiology
Distribution and biosynthesis of cis-9,10-epoxystearic acid in Pneumocystis carinii.
PMID 10519279
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E4765
trans-9,10-Epoxystearic acid, ~99% (capillary GC)
C18H34O3