EMAIL THIS PAGE TO A FRIEND
Amino acids
Effect of some herbicides on the production of lysine by Azotobacter chroococcum.
PMID 10524274
Abstract
Production of lysine by Azotobacter chroococcum strain H23 was studied in chemically-defined media amended with different concentrations of alachlor, metolachlor, 2,4-D, 2,4,5-T and 2,3,6-TBA. The presence of 5, 10, and 50 micrograms/ml of alachlor or 2,3,6-TBA significantly decreased quantitative production of lysine. However, the presence 2,4-D or 2,4,5-T at concentrations of 10 and 50 micrograms/ml enhanced the production of lysine. Quantitative production of lysine was not affected as consequence of the addition of metolachlor to the culture medium, showing that the release lysine to the culture media by A. chroococcum was not affected by that herbicide.