EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health. Part A
3-Methylpentane. CAS# 96-14-0.
PMID 10537375
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68320
3-Methylpentane, analytical standard
C6H14
M66005
3-Methylpentane, ≥99%
C6H14