EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Safety assessment of butylated hydroxyanisole and butylated hydroxytoluene as antioxidant food additives.
PMID 10541460
Abstract
Butylated hydroxyanisole (BHA) and butylated hydroxytoluene (BHT) are widely used antioxidant food additives. They have been extensively studied for potential toxicities. This review details experimental studies of genotoxicity and carcinogenicity which bear on cancer hazard assessment of exposure to humans. We conclude that BHA and BHT pose no cancer hazard and, to the contrary, may be anticarcinogenic at current levels of food additive use.