EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Pesticides in fermentative processes of wine.
PMID 10552734
Abstract
The influence of six fungicides (azoxystrobin, cyprodinil, fludioxonil, mepanipyrim, pyrimethanil, and tetraconazole) on the fermentative activity of two yeasts (Saccharomyces cerevisiae and Kloeckeraapiculata) and two lactic bacteria (Leuconostoc oenos and Lactobacillus plantarum) was studied. The possibility of their being degraded by these yeasts and bacteria was also investigated. The presence of the pesticides did not affect alcoholic fermentation, not even with levels higher than those normally found in grapes in field experiments. On the contrary, their presence stimulated the yeast, especially K. apiculata, to produce more alcohol. The fermentative process did not affect the amount of pesticides either by degradation or by adsorption. During malolactic fermentation by Le. oenos, malic acid decreased slightly less (by approximately 15%) in the presence of all pesticides, except mepanipyrim. A lower effect ( approximately 5%) was found during the fermentative process with La. plantarum. The bacteria studied did not show a degradative effect on pesticides during malolactic fermentation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33970
Mepanipyrim, PESTANAL®, analytical standard
C14H13N3