EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Convenient synthesis of L-proline benzyl ester.
PMID 10560736
Abstract
Mesylates or tosylates of delta-hydroxy-L-norvaline esters spontaneously afford L-proline esters upon exposure to aqueous buffer in near quantitative yield. This novel reaction has led to the development of a simple route to optically active proline esters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364460
L-Proline benzyl ester hydrochloride, 98%
C12H15NO2 · HCl