EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
N-methylation of 1-methyltryptamines by indolethylamine N-methyltransferase.
PMID 1056183
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06840
1-Methyltryptamine, ≥95.0% (GC)
C11H14N2