EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Preferential oxidative dehalogenation upon conversion of 2-halophenols by Rhodococcus opacus 1G.
PMID 10564791
Abstract
The regiospecificity of hydroxylation of C2-halogenated phenols by Rhodococcus opacus 1G was investigated. Oxidative defluorination at the C2 position ortho with respect to the hydroxyl moiety was preferred over hydroxylation at the non-fluorinated C6 position for all 2-fluorophenol compounds studied. Initial hydroxylation of 2,3, 5-trichlorophenol resulted in the exclusive formation of 3, 5-dichlorocatechol. These results indicate that, in contrast to all other phenol ortho-hydroxylases studied so far, phenol hydroxylase from R. opacus 1G is capable of catalyzing preferential oxidative defluorination but also oxidative dechlorination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442285
2,3,5-Trichlorophenol, analytical standard
C6H3Cl3O
290394
2,5-Difluorophenol, 95%
C6H4F2O