EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Absorption, metabolism, and disposition of 1,3-diphenyl-1-triazene in rats and mice after oral, i.v., and dermal administration.
PMID 10570033
Abstract
1,3-Diphenyl-1-triazene (DPT) is used in the synthesis of polymers and dyes, and has been found as an impurity in the color additives D&C Red 33 and FD&C Yellow 5. [(14)C]DPT, randomly labeled in the phenyl rings, was used to investigate its disposition in rodents. Dermal doses to rats and mice (2 and 20 mg/cm(2)) were poorly absorbed (